Skip to Content
Merck
All Photos(1)

Key Documents

38180

Sigma-Aldrich

Diisobutylene

technical, ≥90% (3 parts 2,4,4-trimethyl-1-pentene + 1 part 2,4,4-trimethyl-2-pentene, GC)

Synonym(s):

2,4,4-Trimethyl-1-pentene + 2,4,4-Trimethyl-2-pentene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2C(CH3)=CH2+(CH3)3CCH=C(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1098309
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

technical

Quality Level

Assay

≥90% (3 parts 2,4,4-trimethyl-1-pentene + 1 part 2,4,4-trimethyl-2-pentene, GC)

refractive index

n20/D 1.411

bp

101-103 °C (lit.)

density

0.716 g/mL at 20 °C (lit.)

SMILES string

CC(=C)CC(C)(C)C

InChI

1S/C8H16/c1-7(2)6-8(3,4)5/h1,6H2,2-5H3

InChI key

FXNDIJDIPNCZQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutylene is an acyclic alkene. It is composed of a mixture of 2,4,4-trimethyl-1-pentene and 2,4,4- trimethyl-2-pentene (isomeric forms of diisobutylene). It is isostructural to to iso-octane. It can be synthesized from tertiary butyl alcohol.

Application

Diisobutylene, an alkene, may be used in the following studies:
  • To compose alternalte diesel fuel.
  • As fuel in single-cylinder engine; exhaust generated from combustion due to the burning of fuel has been reported to contain high concentrations of olefins.1
  • Preparation of mono- and dialkylated diphenylamines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The development of a detailed chemical kinetic mechanism for diisobutylene and comparison to shock tube ignition times.
Metcalfe WK, et al.
Proceedings of the Combustion Institute, 31(1), 377-384 (2007)
Alkylation of diphenylamine with α-methylstyrene and diisobutylene using acid-treated clay catalysts.
Chitnis SR and Sharma MM.
J. Catal., 160(1), 84-94 (1996)
The isomers in ?diisobutylene?. II1.
Tongberg CO, et al.
Journal of the American Chemical Society, 54(9), 3706-3710 (1932)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service