Skip to Content
Merck
All Photos(1)

Key Documents

SCT073

Sigma-Aldrich

BioTracker NIR Amyloid-ß Plaque Imaging Dye

Near-Infrared probe for live cell imaging of amyloid-ß (Aß) plaques, critical for the early detection of Alzheimer′s disease.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.21

form

lyophilized (dark red)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

detection method

fluorometric

General description

Alzheimer′s disease (AD) is a progressive neurodegenerative brain disorder and does not have a proper cure yet. The formation and accumulation of amyloid-ß (Aß) plaques in the brain is thought to be a critical pathological hallmark for early diagnosis of AD. Current gold standard probes for histological staining of amyloid fibrils, such as thioflavin derivatives, have several drawbacks. These probes suffer from noises from always-on pattern, limited blood-brain-barrier (BBB) penetration and distorted signals.BioTracker NIR Amyloid-ß Plaque Imaging Dye is aggregation-induced-emission (AIE) active probe that can be used to detect amyloid-ß plaques in vivo as well as ex-vivo. This probe has been tested in mouse models of AD, as well as ex-vivo brain tissue sections. The BioTracker probe possess excellent features, such as: (1) ultrahigh signal-to-noise (S/N) ratio with integrating background minimization and fidelity signal amplification, (2) remarkable binding affinity to Aß plaques with efficient BBB penetrability, and (3) NIR AIE-active emission with excellent photostability. Spectral Properties:Absorbance: 500 nmEmission: 720 nmResearch Category:Alzheimer′s DiseaseLive Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service