Skip to Content
Merck
All Photos(5)

Key Documents

ABN1675M

Sigma-Aldrich

Anti-Nurr-1

from rabbit

Synonym(s):

Nuclear receptor subfamily 4 group A member 2, Immediate-early response protein NOT, Orphan nuclear receptor NURR1, Transcriptionally-inducible nuclear receptor, NR4A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
IHC
WB
species reactivity:
mouse, human
technique(s):
ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NR4A2(4929)

General description

Nuclear receptor subfamily 4 group A member 2 (UniProt: P43354; also known as Immediate-early response protein NOT, Orphan nuclear receptor NURR1, Transcriptionally-inducible nuclear receptor) and is encoded by the NR4A2 (also known as NOT, NURR1, TINUR) gene (Gene ID: 4929) in human. Nurr-1 belongs to the nuclear hormone receptor family that acts as a transcriptional regulator and is considered to be important for the differentiation and maintenance of meso-diencephalic dopaminergic (mdDA) neurons during development. It is also crucial for expression of a set of genes such as SLC6A3, SLC18A2, TH and DRD2 which are essential for development of mdDA neurons. It is expressed in a number of T cell, B cell and fibroblast cell lines, but its expression is higher in brain tissue. Two isoforms of Nurr-1 have been reported that are produced by alternative splicing. The ligand-binding domain (LBD) of Nurr-1 contains no cavity as a result of the tight packing of side chains from several bulky hydrophobic residues in the region normally occupied by ligands. It also lacks a classical binding site for co-activators. However, it has hydrophobic region on the surface of LBD, which is critical for its activity. (Ref.: Volakakis N et al (2006). J. Mol. Endocrinol 37(2); 317-326; Wang G et al. (2003). Nature 423(6939):555-560).

Specificity

This rabbit polyclonal antibody detects Nurr-1 in human and murine tissues. It targets an epitope within the C-terminal half.

Immunogen

Epitope: domain of:
His-tagged recombinant fragment corresponding to 271 amino acids from the C-terminal half of human Nurr-1 and includes the ligand binding domain.

Application

Anti-Nurr-1, Cat. No. ABN1675, is a highly specific rabbit polyclonal antibody that targets Nuclear receptor subfamily 4 group A member 2 and has been tested in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Nurr-1 in mouse hippocampus and human cerebellum tissue sections.

ELISA Analysis: A representative lot detected recombinant Nurr-1 LBDs (Courtesy of Dr. Pierre Leblanc at McLean Hospital, Belmont, MA).

Immunohistochemistry Analysis: A representative lot detected Nurr-1 in Midbrain dopamine neurons (Leblanc, P., et. al. (2015). J Vis Exp. (102):e52963).

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected Nurr-1 in HEK293T and CHO cells overexpressed with Myc-Tagged mouse full length Nurr1/Nor1/Nurr77 (Courtesy of Dr. Pierre Leblanc at McLean Hospital, Belmont, MA).

Immunohistochemistry Analysis: A 1:250-1:2,500 dilution from a representative lot detected Nurr-1 in mouse brain tissue sections.(Courtesy of Dr. Pierre Leblanc at McLean Hospital, Belmont, MA).

ELISA Analysis: A representative lot detected Nurr-1 ligand binding domain (Leblanc, P., et. al. (2015). J Vis Exp. (102):e52963).

Western Blotting Analysis: A representative lot detected Nurr-1 in CHO cells transfected with full-length myc-tagged Nurr-1 (Leblanc, P., et. al. (2015). J Vis Exp. (102):e52963).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in mouse cerebral cortex tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Nurr-1 in mouse cerebral cortex tissue sections.

Target description

~70 kDa observed; 66.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum in PBS with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service