Skip to Content
Merck
All Photos(2)

Key Documents

AB15928

Sigma-Aldrich

Anti-Vitamin K dependent protein S Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

protein S (alpha), protein S, alpha, protein Sa, vitamin K-dependent plasma protein S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, rhesus macaque, monkey

species reactivity (predicted by homology)

horse, chimpanzee, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PROS1(5627)

General description

Vitamin K dependent protein S is an anticoagulant plasma protein; it is a cofactor to activated protein C in the degradation of coagulation factors Va and VIIIa. It helps to prevent coagulation and stimulating fibrinolysis.

Specificity

Monkey (93%) and Rat (93%) Rhesus Monkey (93%).
This antibody recognizes Vitamin K dependent protein S.

Immunogen

This antibody is generated from rabbits immunized with a KLH-conjugated linear peptide corresponding to residues surrounding the mature peptide region of Vitamin K dependent protein S.

Application

Detect Vitamin K dependent protein S using this Anti-Vitamin K dependent protein S Antibody validated for use in WB.
Immunoprecipitation Analysis: 1:50,000 dilution from a previous lot detected Vitamin K dependent protein S on 15 µg of normal mouse serum.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in mouse liver tissue lysate.
Western Blot Analysis: : 1:1000 dilution of this antibody detected Vitamin K dependent protein S on 10 µg of mouse liver tissue lysate.

Target description

Approx. 69-75 kDa

Physical form

Affinity purified
Purified rabbit polyclonal containing 0.1 M Tris-glycine, pH 7.4, 0.150 mM NaCl and 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
Mouse liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Lumbroso et al.
Frontiers in immunology, 9, 358-358 (2018-03-17)
The complete resolution of inflammation requires the uptake of apoptotic polymorphonuclear cells (PMN) by local macrophages (efferocytosis) and the consequent reprogramming of the engulfing phagocytes to reparative and pro-resolving phenotypes. The tyrosine kinase receptors TYRO3, AXL, and MERTK (collectively named
Lack of protein S in mice causes embryonic lethal coagulopathy and vascular dysgenesis.
Burstyn-Cohen, T; Heeb, MJ; Lemke, G
The Journal of Clinical Investigation null
Genetic dissection of TAM receptor-ligand interaction in retinal pigment epithelial cell phagocytosis.
Burstyn-Cohen, T; Lew, ED; Traves, PG; Burrola, PG; Hash, JC; Lemke, G
Neuron null
Maria Nassar et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(3), E337-E346 (2017-01-05)
The oral epithelium contributes to innate immunity and oral mucosal homeostasis, which is critical for preventing local inflammation and the associated adverse systemic conditions. Nevertheless, the mechanisms by which the oral epithelium maintains homeostasis are poorly understood. Here, we studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service