8.00202
Ethyl octanoate
for synthesis
Synonym(s):
Ethyl octanoate, Ethyl caprylate, Octanoic acid ethyl ester
About This Item
Recommended Products
vapor pressure
0.2 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
liquid
autoignition temp.
325 °C
potency
25960 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)
expl. lim.
0.7 % (v/v)
bp
208 °C/1013 hPa
mp
-47 °C
transition temp
flash point 79 °C
density
0.87 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CC)C(=O)CCCCCCC
InChI
1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3
InChI key
YYZUSRORWSJGET-UHFFFAOYSA-N
Related Categories
Analysis Note
Density (d 20 °C/ 4 °C): 0.865 - 0.867
Identity (IR): passes test
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
167.0 °F - closed cup
Flash Point(C)
75 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service