Skip to Content
Merck
All Photos(1)

Documents

524403

Sigma-Aldrich

Pravastatin, Sodium Salt

A water-soluble, competitive 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that potently blocks in vivo cholesterol synthesis (Ki = ~1 nM).

Synonym(s):

Pravastatin, Sodium Salt, (βR,δR,1S,2S,6S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,δ,6-trihydroxy-2-methyl-8[(2S)-2-methyl-1-oxobutoxyl]-1-naphthaleneheptanoic Acid Na, 3β-Hydroxycompactin, Na

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C23H36O7.Na/c1-4-13(2)23(29)30-20-11-17(25)9-15-6-5-14(3)19(22(15)20)8-7-16(24)10-18(26)12-21(27)28;/h5-6,9,13-14,16-20,22,24-26H,4,7-8,10-12H2,1-3H3,(H,27,28);/q;+1/p-1/t13-,14-,16+,17+,18+,19-,20-,22-;/m0./s1

InChI key

VWBQYTRBTXKKOG-IYNICTALSA-M

General description

A water-soluble, competitive 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that blocks in vivo cholesterol synthesis (Ki ~1 nM). Also acts as an immunomodulator and an inhibitor of ras p21 isoprenylation.
A water-soluble, competitive 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that potently blocks in vivo cholesterol synthesis (Ki = ~1 nM). Offers cardioprotection. Also acts as an immunomodulator and an inhibitor of ras p21 isoprenylation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
AMG-CoA reductase
Product does not compete with ATP.
Reversible: no
Target Ki: ~1 nM inhibiting 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase and blocking in vivo cholesterol synthesis

Packaging

Packaged under inert gas

Warning

Toxicity: Environmental Hazard (N)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Kwak, B., et al. 2000. Natl. Med.6, 1399.
Hamelin, B.A., and Turgeon, J., 1998. Trends Pharmacol. Sci.19, 26.
Narisawa, T., et al. 1996. Jpn. J. Cancer Res.87, 798.
Cohen, L.H., et al. 1993. Biochem. Pharmacol.45, 2203.
Koga, T., et al. 1990. Biochim. Biophys. Acta.1045, 115.
Yoshino, G., et al. 1986. Diabetes Res. Clin. Pract. 2, 179

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service