Skip to Content
Merck
All Photos(1)

Key Documents

5.08148

Sigma-Aldrich

Nefazodone Hydrochloride

Synonym(s):

Nefazodone Hydrochloride, 5-HT 2A Serotonin Receptor Antagonist, Nefazodone, BMY-13754, MJ-13754-1, Serzone, 5-HT2A Serotonin Receptor Antagonist, Nefazodone, BMY-13754, MJ-13754-1, Serzone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32ClN5O2 · xHCl
CAS Number:
Molecular Weight:
470.01 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

26 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 10 mM
DMSO: 25 mM

storage temp.

2-8°C

SMILES string

Clc1cc(ccc1)N2CCN(CC2)CCC[n]3nc([n]([c]3=O)CCOc4ccccc4)CC.Cl

InChI

1S/C25H32ClN5O2.ClH/c1-2-24-27-31(25(32)30(24)18-19-33-23-10-4-3-5-11-23)13-7-12-28-14-16-29(17-15-28)22-9-6-8-21(26)20-22;/h3-6,8-11,20H,2,7,12-19H2,1H3;1H

InChI key

DYCKFEBIOUQECE-UHFFFAOYSA-N

General description

A potent 5-HT2A receptor antagonist (Kd = 26 nM), with moderate affinity for the alpha1-adrenergic receptor (Kd = 48 nM) and 5-HT1A receptor (Kd = 80 nM). Originally developed as a clinical antidepressant but discontinued in 2004, nefazodone (Serzone) still remains a specific inhibitor of certain serotonergic pathways. Nefazodone also has been reported to induce mitochondrial dysfuntion.

Warning

Toxicity: Standard Handling (A)

Other Notes

Kim, J.-A. et al. 2012. Toxicol. Lett.214, 175.
Cusack, B. et al. 1994. Psychopharmacol.114, 559.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service