Skip to Content
Merck
All Photos(1)

Key Documents

1.02216

Supelco

Charcoal activated

for the determination of AOX

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522
EC Index Number:
231-153-3
NACRES:
SE.11
Pricing and availability is not currently available.

Quality Level

form

solid

technique(s)

photometry: suitable

mp

3550 °C

bulk density

150‑440 kg/m3

storage temp.

2-30°C

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Analysis Note

Iodine number: ≥ 1050 mg/g
AOX (as chloride): ≤ 0.0015 %
Particle size (50 - 150 µm): ≥ 80 %
Specific surface area (BET): about 800

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service