1.01209
Ammonium sulfate
99.9999 Suprapur®
Synonym(s):
Mascagnite
About This Item
Recommended Products
vapor pressure
<1 Pa ( 25 °C)
Quality Level
description
for inorganic trace analysis
Assay
99.9999%
form
solid
potency
2840 mg/kg LD50, oral (Rat)
pH
5 (100 g/L in H2O)
mp
>280 °C (dec.) (lit.)
solubility
754-764 g/L
density
1.77 g/mL at 25 °C (lit.)
bulk density
850 kg/m3
storage temp.
2-30°C
SMILES string
N.N.OS(O)(=O)=O
InChI
1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)
InChI key
BFNBIHQBYMNNAN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Isolation and identification of milk-clotting proteases from Prinsepia utilis Royle and its application in cheese processing.: This study utilized ammonium sulfate for the isolation of milk-clotting proteases, which were then applied in cheese processing, enhancing the efficiency and quality of cheese production (Zhang et al., 2024).
- Diffusion-driven fed-batch fermentation in perforated ring flasks.: The research used ammonium sulfate in fed-batch fermentation processes to enhance microbial growth and product yield, demonstrating its utility in optimizing industrial fermentation techniques (Lüchtrath et al., 2024).
- Ammonium Sulfate to Modulate Crystallization for High-Performance Rigid and Flexible Perovskite Solar Cells.: This research investigates the role of ammonium sulfate in modulating the crystallization process of perovskite solar cells, significantly improving their performance and stability (Zou et al., 2024).
- Effectiveness of three nitrification inhibitors on mitigating trace gas emissions from different soil textures under surface and subsurface drip irrigation.: The study utilized ammonium sulfate in combination with nitrification inhibitors to reduce trace gas emissions in agricultural settings, highlighting its environmental benefits (Ribeiro et al., 2024).
Analysis Note
Assay (acidimetric): ≥ 99.5 %
Chloride (Cl): ≤ 3 ppm
Nitrate (NO₃): ≤ 10 ppm
Phosphate (PO₄): ≤ 5 ppm
Ag (Silver): ≤ 0.010 ppm
Al (Aluminium): ≤ 0.050 ppm
Ba (Barium): ≤ 0.050 ppm
Be (Beryllium): ≤ 0.010 ppm
Bi (Bismuth): ≤ 0.050 ppm
Ca (Calcium): ≤ 0.100 ppm
Cd (Cadmium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.005 ppm
Fe (Iron): ≤ 0.050 ppm
Ga (Gallium): ≤ 0.050 ppm
In (Indium): ≤ 0.050 ppm
K (Potassium): ≤ 0.100 ppm
Li (Lithium): ≤ 0.050 ppm
Mg (Magnesium): ≤ 0.010 ppm
Mn (Manganese): ≤ 0.010 ppm
Na (Sodium): ≤ 0.100 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.010 ppm
Tl (Thallium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.010 ppm
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service