Skip to Content
Merck
All Photos(1)

Key Documents

810299P

Avanti

25-(C4 TopFluor) 25-OH cholesterol

Avanti Research - A Croda Brand 810299P, powder

Synonym(s):

25-[4-(dipyrrometheneboron difluoride)butanoyl]-25-hydroxycholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H65 BF2N2O3
CAS Number:
Molecular Weight:
718.81
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810299P-1MG)
pkg of 1 × 100 μg (810299P-100ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810299P

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (810299P-1MG)
5 mL Amber Glass Screw Cap Vial (810299P-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service