Key Documents
Z278432
Ace replacement cap
with hole
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
packaging
pk of 48 ea
manufacturer/tradename
Ace Glass 9590-46
General description
Caps with hole fit threaded outer 14/10 joints and, with an O-ring (Z504874), form an "O-RING-CAP SEAL" connection. Size: 20 mm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)