Key Documents
Z180629
Ace European-style three-neck flask
capacity 250 mL
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flask
three-neck
Joint
center joint ST/NS29/42
side joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6961-40
capacity
250 mL
General description
Has tapered walls with shallow bottom for more efficient magnetic or mechanical stirring with half-moon shaped paddle stirrer. 7 mm O.D. threaded thermometer port with nylon bushing and FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)