Skip to Content
Merck
All Photos(1)

Key Documents

W205001

Sigma-Aldrich

Ambrette seed absolute

FG

Synonym(s):

Hibiscus abelmoschus l. seed absolute

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2050
Council of Europe no.:
228n
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Kosher
biological source:
Hibiscus abelmoschus
food allergen:
no known allergens

biological source

Hibiscus abelmoschus

Quality Level

300
400

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.475 (lit.)

bp

298 °C/755 mmHg (lit.)

density

0.9 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; woody; tobacco

Preparation Note

Extraction method: steam distillation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service