Skip to Content
Merck
All Photos(1)

Documents

GF91691983

Osmium

powder, 500 max. part. size (micron), weight 10 g, purity 99.9%

Synonym(s):

OS006010

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141732
NACRES:
NA.23

Assay

99.9%

form

powder

manufacturer/tradename

Goodfellow 916-919-83

weight

10 g

max. part. size

500 (micron)

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service