Skip to Content
Merck
All Photos(1)

Documents

901716

Sigma-Aldrich

Sucrose-(fructose-13C6)

endotoxin tested, ≥99 atom % 13C, ≥99% (CP)

Synonym(s):

D-(+)-Saccharose-(fructose-13C6), Sugar-(fructuose-13C6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C6H22O11
CAS Number:
Molecular Weight:
348.25
MDL number:
UNSPSC Code:
12352201

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

solid

mp

185-187 °C (lit.)

suitability

endotoxin tested

mass shift

M+6

SMILES string

OC[C@H]1O[C@H](O[13C@]2([13CH2]O)O[13C@H]([13CH2]O)[13C@@H](O)[13C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the enduser.


Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service