Skip to Content
Merck
All Photos(1)

Key Documents

809640

Sigma-Aldrich

Dehydroepiandrosterone-2,2,3,4,4-d5

≥98 atom % D, ≥97% (CP)

Synonym(s):

DHEA-2,2,3,4,4-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D5H23O2
CAS Number:
Molecular Weight:
293.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

O=C1CCC2C3CC=C4C([2H])([2H])[C@@]([2H])(O)C([2H])([2H])C[C@]4(C)C3CC[C@@]21C

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14?,15?,16?,18-,19-/m0/s1/i7D2,11D2,13D

InChI key

FMGSKLZLMKYGDP-WJKQEKIRSA-N

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saé Muñiz-Hernández et al.
Microorganisms, 9(3) (2021-04-04)
Toxoplasmosis is a zoonotic disease caused by the apicomplexa protozoan parasite Toxoplasma gondii. This disease is a health burden, mainly in pregnant women and immunocompromised individuals. Dehydroepiandrosterone (DHEA) has proved to be an important molecule that could drive resistance against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service