Skip to Content
Merck
All Photos(1)

Key Documents

756962

Sigma-Aldrich

3-Bromooxetane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BrO
CAS Number:
Molecular Weight:
136.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.484

density

1.651 g/mL at 25 °C

functional group

bromo
ether

storage temp.

−20°C

SMILES string

BrC1COC1

InChI

1S/C3H5BrO/c4-3-1-5-2-3/h3H,1-2H2

InChI key

SZTIZZFKWQWSSP-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service