618535
Water-17O
80-84.9 atom % 17O
Synonym(s):
17O Labeled water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
isotopic purity
80-84.9 atom % 17O
mol wt
18.82 by atom % calculation
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.044 g/mL at 25 °C
bulk density
1.01
mass shift
M+1
SMILES string
[17OH2]
InChI
1S/H2O/h1H2/i1+1
InChI key
XLYOFNOQVPJJNP-OUBTZVSYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service