Merck
  • Catalytic asymmetric synthesis of butane diacetal-protected (4S,5S)-dihydroxycyclohexen-1-one and use in natural product synthesis.
Catalytic asymmetric synthesis of butane diacetal-protected (4S,5S)-dihydroxycyclohexen-1-one and use in natural product synthesis.
Organic & biomolecular chemistry (2012-08-30)
David J Burns, Shuji Hachisu, Peter O'Brien, Richard J K Taylor
ABSTRACT
Due to the lack of availability of unnatural (+)-quinic acid as a starting material, a 6-step synthesis of butane diacetal-protected (4S,5S)-dihydroxycyclohexen-1-one (formally derived from (+)-quinic acid) has been devised. The key catalytic asymmetric step involves a chiral Co-salen-catalysed epoxide ring-opening reaction. (4S,5S)-Dihydroxycyclohexen-1-one was utilised in the synthesis of two cyclohexenone natural products isolated from the mycelia of Lasiodiplodia theobromae.
Sigma-Aldrich
N,N′-Bis(salicylidene)ethylenediamine, 98%