Merck
  • IgE antibodies against piperazine and N-methyl-piperazine in two asthmatic subjects.
IgE antibodies against piperazine and N-methyl-piperazine in two asthmatic subjects.
International archives of allergy and applied immunology (1986-01-01)
H Welinder, L Hagmar, C Gustavsson
ABSTRACT
The presence of specific IgE antibodies against conjugates between human serum albumin and piperazine, a compound previously known to cause occupational asthma, and N-methyl-piperazine, respectively, was demonstrated by RAST, RAST inhibition and skin prick tests in 2 asthmatic subjects occupationally exposed to these agents.
Sigma-Aldrich
1-Methylpiperazine, 99%
Pricing and availability is not currently available.
Supelco
1-Methylpiperazine, analytical standard
Pricing and availability is not currently available.