Skip to Content
Merck
All Photos(1)

Key Documents

48081

Supelco

Perylene-d12 solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 525,525.1,525.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c21c3c4c5c(c2c(c(c(c1c(c(c3[2H])[2H])[2H])[2H])[2H])[2H])c(c(c(c5c(c(c4[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

CSHWQDPOILHKBI-AQZSQYOVSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ou-Ya Zhao et al.
Chemosphere, 174, 774-780 (2017-02-16)
The present study used strain ZH-H2 (Fusarium sp.) isolated by our group as the PAH-degrading strain and 5-6-rings PAHs as degradation objects. The soil incubation experiment was carried out to investigate the starch-enhanced degradation effects of HMW PAHs by Fusarium
Xiaoxue Zhang et al.
Environmental science and pollution research international, 27(34), 42969-42978 (2020-07-30)
The capacity of Fusarium sp. strain ZH-H2 to secret lignin peroxidase (LiP), laccase (Lac), and manganese peroxidase (MnP) and degrade high molecular weight polycyclic aromatic hydrocarbons (HMW-PAHs) was studied. When the fungus was grown in control mineral salt medium for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service