Key Documents
2636801
Supelco
Inlet Liner, Splitless Type, Single Taper Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
12000000
material
glass liner
description
Universal flash
feature
glass flash insert
packaging
pkg of 1 ea
manufacturer/tradename
Varian 03-918339-00 (OEM)
L × O.D. × I.D.
73 mm × 6.35 mm × 4.0 mm
compatibility
for use with Varian 1075/1077 Injectors
technique used for 0.53 mm I.D. columns
Related Categories
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Other Notes
I263680 --> material nos. 2636801 (1pk), 2636805 (5pk), & 2636825 (25pk).
Choose from one of the most recent versions:
Certificates of Analysis (COA)