Skip to Content
Merck
All Photos(1)

Key Documents

Z756717

Clear view conical tube holder

Holds 1 x 15 or 50 mL tube, white polypropylene, autoclavable

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.22

material

white polypropylene

feature

autoclavable
Holds 1 x 15 or 50 mL tube

packaging

bag of 4 ea

manufacturer/tradename

Bel-Art F18796-0000

General description

The most versatile tube holder on the market. Set-up takes just seconds and gives you the choice of creating a 15 mL or a 50 mL tube holder.

  • Open uprights allow viewing of the entire contents of the tube
  • Tube can be easily lifted from the holder, or pinch the uprights and carry the holder with the tube
  • Autoclavable polypropylene

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service