Z740296
RotoFlex® Plus bench top tube rotator rotisserie
for 50 mL tubes/6 tube capacity per rotisserie
All Photos(1)
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Cole Parmer 439739
General description
The RotoFlex will accommodate a variety of tubes from 0.5 - 50 mL by simply changing the rotisserie.
Legal Information
RotoFlex is a registered trademark of Argos Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)