Skip to Content
Merck
All Photos(2)

Key Documents

Z722375

IKA® ULTRA-TURRAX® disperser tubes

DT-20 dispersing tube with rotor-stator element, 25/cs

Synonym(s):

tissue grinders, tissue homogenizers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.

packaging

pack of 25

manufacturer/tradename

IKA 3703100

capacity

5-15 mL

Looking for similar products? Visit Product Comparison Guide

General description

IKA® ULTRA-TURRAX® Disperser Tubes.

These hermetically sealed tubes prevent the spreading of aerosols or vapours created by mechanical stirring, dispersion or grinding infectious, toxic or odourous substances in open vessels. Disposable and single-use tubes, they avoid the possibility of cross-contamination of samples and remove the need for clean up of disperser tools, stirrer tools, grinding chambers or vessels.

Each tube has a bayonet fitting on the bottom, for easy, secure connection to the tube drive.

Features and Benefits

  • Includes tube, dispersing device, cover
  • Volume: 5 - 15 ml
  • Application: Dispersion, homogenisation, suspensions, pharmacokinetics, metabolism studies, diagnosis
  • Additional media quantities may be added during operation

Warning

Stirring, dispersing or grinding may generate heat or cause other changes in the sample that might lead to build up of pressure inside the tube. Use appropriate personal protective equipment when operating and opening tubes.

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service