VIV102008
SynVivo SynRAM 3D Inflammation model chip, IMN2 radial configuration
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
60104506
NACRES:
NA.84
Recommended Products
packaging
case of 1 ea
manufacturer/tradename
SynVivo 102008
storage temp.
room temp
General description
SynRAM Inflammation model chip - Barrier type: Pillars, Distance between pillars: 8 μm, Barrier height: 3 μm, Travel/barrier width: 50 μm, Outer Channel width: 200 μm, Device depth: 100 μm. This chip is used to create the SynRAM inflammation model in monoculture or co-culture configurations
Linkage
LInk to Publications
application text
application text
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service