Skip to Content
Merck
All Photos(1)

Key Documents

SML3082

Sigma-Aldrich

Demethylwedelolactone

≥98% (HPLC)

Synonym(s):

1,3,8,9-Tetrahydroxy-6H-benzofuro[3,2-c][1]benzopyran-6-one, 1,3,8,9-Tetrahydroxy-6H-benzofuro[3,2-c]chromen-6-one; 1,3,8,9-Tetrahydroxycoumestan, Desmethylwedelolactone, Norwedelolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8O7
CAS Number:
Molecular Weight:
300.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to green

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1OC2=C(C(O)=CC(O)=C2)C3=C1C4=C(O3)C=C(O)C(O)=C4

InChI

1S/C15H8O7/c16-5-1-9(19)13-11(2-5)22-15(20)12-6-3-7(17)8(18)4-10(6)21-14(12)13/h1-4,16-19H

InChI key

LUTYTNLPIUCKBJ-UHFFFAOYSA-N

Biochem/physiol Actions

Demethylwedelolactone is a coumestan extracted from Eclipta alba that exhibits numerous biological activities including anti-inflammatory and hepato-protective. Demethylwedelolactone inhibits MDA-MB-231 breast cancer cells motility and invasiveness. It suppresses the metastasis and lung colonization of the tumor cells in the nude mice. Both demethylwedelolactone and wedelolactone appear to attenuate cigarette smoking extract induced inflammation through blockade of autophagy flux in respiratory epithelial cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Satheesh Naik et al.
Maedica, 13(3), 217-222 (2019-09-07)
The hepatic injury due to oxidative stress was ameliorated through administration of an aqueous extract of Eclipta alba leaves and suggested that wedelolactone and demethylwedelolactone were the possible components of Eclipta alba behind the protective effect on liver as well
Shumin Ding et al.
Frontiers in pharmacology, 9, 107-107 (2018-03-09)
Excessive autophagy plays a crucial role in cigarette smoking extract (CSE)-induced inflammation response and oxidative damage of respiratory epithelial cells. The components from Eclipta prostrata L. (CCE) have been shown to be beneficial for CSE-induced epithelial cells injury. However, whether
Yean-Jang Lee et al.
European journal of medicinal chemistry, 56, 361-367 (2012-08-29)
The anticancer properties of demethylwedelolactone (DWEL) and wedelolactone (WEL), which are naturally occurring coumestans, have not been well characterized. In this study, we investigated the anti-invasive effects of synthetic WEL and DWEL on human MDA-MB-231 breast cancer cells. We found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service