Skip to Content
Merck
All Photos(4)

Key Documents

SAB5701193

Sigma-Aldrich

Anti- CDKN1B/p27KIP1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).

To order products, please contact your local dealer.

Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€197.00


Availability
Usually ships in 3 weeks (4 to 6 weeks for custom orders).

To order products, please contact your local dealer.

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

mouse, human, rat

concentration

0.43 mg/mL

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU127601EHU084601EHU067721
Ensembl | human accession no.

ENSG00000025039

Ensembl | human accession no.

ENSG00000011451

Ensembl | human accession no.

ENSG00000165175

Ensembl | human accession no.

ENSG00000112640

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

human ... RRAGD(58528), RRAGD(58528)

Gene Information

human ... WIZ(58525), WIZ(58525)

Gene Information

human ... MID1IP1(58526), MID1IP1(58526)

Gene Information

human ... PPP2R5D(5528), PPP2R5D(5528)

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

This gene encodes a cyclin-dependent kinase inhibitor, which shares a limited similarity with CDK inhibitor CDKN1A/p21. The encoded protein binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. Mutations in this gene are associated with multiple endocrine neoplasia type IV (MEN4). [provided by RefSeq, Apr 2014]

Immunogen

A synthesized peptide derived from human CDKN1B/p27KIP1

Physical form

Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service