Skip to Content
Merck
All Photos(1)

Key Documents

H0413

Millipore

HIS-Select® Filter Plate

Synonym(s):

His-tag filtered microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

grade

Molecular Biology
for molecular biology

form

plate

suitability

suitable for (cell culture)

storage temp.

2-8°C

General description

The HIS-Select® Filter Plate is an immobilized metal-ion affinity chromatography (IMAC) product that allows rapid purification of small-scale crude cell extracts containing histidine-tagged proteins. The HIS-Select® Filter Plate contains 20 μm, spherical silica particles (100 nm pore size) with a hydrophilic layer.

The HIS-Select® Filter Plate is selective for recombinant proteins with histidine tags and exhibits very low non-specific binding of other proteins. The selectivity can be modulated with the inclusion of imidazole during chromatography. The binding capacity of the HIS-Select® Filter Plate is typically > 2 mg per well as determined with an approximately 30 kDa histidine-tagged protein.

Application

The HIS-Select® Filter Plate is suitable for purification of recombinant proteins with histidine tags.

Features and Benefits

  • High Selectivity for his-tagged proteins
  • Ready-to-use plate format
  • Compatible with CelLytic cell lysis reagents

Legal Information

Covered by US Patent No 6,623,655 and are sold for research use only. Commercial use requires additional licenses.
CelLytic is a trademark of Sigma-Aldrich Co. LLC
HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin D Allen et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(46), 19838-19843 (2010-11-04)
The stability, activity, and solubility of a protein sequence are determined by a delicate balance of molecular interactions in a variety of conformational states. Even so, most computational protein design methods model sequences in the context of a single native
Isabel K Macdonald et al.
PloS one, 7(7), e40759-e40759 (2012-07-21)
An assay employing a panel of tumor-associated antigens has been validated and is available commercially (EarlyCDT®-Lung) to aid the early detection of lung cancer by measurement of serum autoantibodies. The high throughput (HTP) strategy described herein was pursued to identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service