Skip to Content
Merck
All Photos(1)

Key Documents

GESE282N2

GE alumina plate

notched, L × D 10 cm × 12 cm

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
JP¥11,100
250 ML
JP¥37,300
1 L
JP¥157,000

JP¥11,100


Availability

Available to ship on06 January 2025Details


To order products, please contact your local dealer.

Select a Size

Change View
50 ML
JP¥11,100
250 ML
JP¥37,300
1 L
JP¥157,000

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

JP¥11,100


Availability

Available to ship on06 January 2025Details


To order products, please contact your local dealer.

packaging

pack of 2 ea

manufacturer/tradename

Cytiva Article No. 80-6153-15
Cytiva SE282N-2

L × D

10 cm × 12 cm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
RAB1372RAB1374RAB1161
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type serum sample(s)
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.22 ng/mL
standard curve range: 1.229-300 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.082 ng/ml

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service