EP4924000061
Eppendorf® Reference® 2 G
single-channel, variable, volume 20-200 μL, yellow, includes epT.I.P.S.® box
Synonym(s):
Adjustable volume pipette
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
feature
includes epT.I.P.S.® box
single-channel, variable
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4924000061
volume
20-200 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Extraordinary precision and accuracy – Eppendorf´s premium pipette
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
Besides improved ergonomics the design focus lays on providing the most accurate results possible, optimum user safety, reliability and robustness for a long service life. The innovative single button operation is not only fast and easy, but it also allows active aerosol1) reduction and thus protects the user, the sample and the pipette. The sturdy handle manufactured with stainless steel protects the pipette from impact and in combination with the spring loaded bumpers the Reference 2 is likely to stay within calibration even if it takes a fall. These high levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducibility, Eppendorf′s Reference 2 is an ideal instrument for these liquid handling applications.
Single-Button Operation
- Lower Operating forces for aspiration/dispensing/tip ejection limit strain on the user.
- Allows users to move more quickly through their pipetting tasks by not having to move the thumb from side to side to eject tips.
- With the ability to eject a tip immediately following dispensing, aerosol contamination of your pipette is considerably reduced.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service