EP0030133374
Eppendorf twin.tec® PCR Plates 96
clear plate, divisible, unskirted, conical bottom colorless wells, capacity 250 μL, colorless, unskirted, divisible, pkg of 20 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear plate
conical bottom colorless wells
conical bottom colorless wells
size:
96 wells
feature:
skirt: no
divisible, unskirted
divisible, unskirted
Recommended Products
material
clear plate
conical bottom colorless wells
feature
skirt: no
divisible, unskirted
packaging
pkg of 20 ea
manufacturer/tradename
Eppendorf® 0030133374
capacity
250 μL
size
96 wells
color
colorless
Looking for similar products? Visit Product Comparison Guide
General description
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Application
PCR
Features and Benefits
- Maximum recovery rates for improved assay results
- Available with barcode (upon request)
- Ideal for quantitative real-time PCR as well
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack® matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service