htr-9c cell line
10080902, Mouse monoclonal against human CD-120a
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Human x Mouse Hybridoma
ImmunoDon
Mouse (Mus musculus) spleen
Myeloma
PAI
Reactivity
Human CD120a (TNF receptor p55)
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Not specified
Cell Line Description
Mouse monoclonal against human CD-120a. TNF receptor p55 prepared from HL-60 cells (htr-mAb series).
Immunogen
Human CD120a (TNF receptor p55)
Application
FACS, Sandwich ELISA for soluble p55 TNF-R.
Culture Medium
Iscoves Modified Dulbeccos Medium (IMDM) + 2 mM glutamine + 25 U/ml Interleukin-6 (IL-6) + 5-10% Fetal Bovine Serum (FBS). Where the cells are grown under static conditions, 0.02% EDTA maybe required to dislodge them from the plastic.
Subculture Routine
Seed cultures between 1-3 x 105 cells/ml; Saturation density 8-10 x 105 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service