AFRC MAC 252
Reactivity: Anti-Abscisic acid, plant growth hormone, 01071008
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Rat x Rat Hybridoma
ImmunoDon
LOU rat spleen
Myeloma
Y3 Ag 1.2.3
Reactivity
Anti-Abscisic acid, plant growth hormone
technique(s)
cell culture | mammalian: suitable
isotype
IgG2a
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Y3Ag1.2.3 x Rat LOU spleen
Cell Line Description
This hybridoma is a subclone of AFRC MAC 60 (86052303). The antibody is directed against the plant growth substance cis, trans-abscisic acid. The rate of antibody production has been tested at ECACC. It must be noted that this hybridoma produces less than 2 μg/ml of purified antibody.
Immunogen
S-cis, trans-abscisic acid - 4(1) - p-amino-benzoyl hydrazone-KLH conjugate
Culture Medium
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2, 37°C. Require mouse macrophage feeder layers upon resuscitation.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service