Key Documents
AXYP96450RC
Round bottom assay plate
size 96 wells, round bottom clear polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs
Synonym(s):
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear plate
round bottom clear polypropylene wells
maximum volume:
500 μL
size:
96 wells
working volume:
450 μL
material
clear plate
round bottom clear polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc P-96-450R-C
Corning
packaging
pkg of 10x5plates/cs
maximum volume
500 μL
size
96 wells
working volume
450 μL
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)