Skip to Content
Merck
All Photos(1)

Key Documents

AXYMCT200CS

Microtubes

size 2.0 mL, clear, pkg of 10x250tubes/cs, sterile

Synonym(s):

Corning tubes, cap tubes, polypropylene tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22
Pricing and availability is not currently available.

material

cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap

sterility

sterile

feature

RNase and DNase free
endotoxin free
graduations

manufacturer/tradename

Axygen Inc MCT-200-C-S
Corning

packaging

pkg of 10x250tubes/cs

size

2.0 mL

color

clear
clear

fitting

cap (push fit)

suitability

suitable for (centrifugation)

Looking for similar products? Visit Product Comparison Guide

General description

  • MaxyClear microtubes are manufactured using virgin polypropylene.
  • Available with a snap cap that ensures an efficient seal for sample protection.
  • Frosted surface for labeling.
  • A piercing port in the center of the cap allows easy access to samples by syringes.
  • Free of RNase/DNase and nonpyrogenic.
  • Maximum G-force is 14,000 RCF
  • Certified boil-proof.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service