PHR2876
Isoflurane Related Compound B
pharmaceutical secondary standard, certified reference material
Synonym(s):
Difluoromethyl 2,2,2-trifluoroethyl ether, 2,2,2-Trifluoroethyl difluoromethyl ether
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1349025
API family
isoflurane
CofA
current certificate can be downloaded
packaging
pkg of 0.5 mL
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
FC(F)(F)COC(F)F
InChI
1S/C3H3F5O/c4-2(5)9-1-3(6,7)8/h2H,1H2
InChI key
ZASBKNPRLPFSCA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT SE 3
Target Organs
Central nervous system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service