Skip to Content
Merck
All Photos(1)

Key Documents

NIST1256B

Aluminum alloy 380

NIST® SRM® 1256b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352311
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1256b comprises a single disk measuring approximately 6.3 cm in diameter and 1.9 cm in thickness. For more information, please refer to the SDS and COA.

SRM 1256B_cert

SRM 1256B_SDS

Application

The Standard Reference Material (SRM) is primarily intended for the evaluation of instrumental analytical methods, such as glow discharge optical emission spectrometry, spark source optical emission spectrometry, and X-ray fluorescence spectrometry.

Features and Benefits

  • NIST certified values and reference values for constituents in aluminum alloy are reported as mass fraction.
  • Accompanied by expiration details of the SRM certification and instructions for use.

Other Notes

Certified for the analytes listed below.
Silicon (Si), Iron (Fe), Copper (Cu), Manganese (Mn), Magnesium (Mg), Nickel (Ni), Zinc (Zn), Titanium B (Ti), Titanium C (Ti), Vanadium B (V), Vanadium C (V), Chromium (Cr), Strontium B (Sr), Strontium C (Sr), Lead (Pb)

Reference/Informational value is provided for the following analytes. Gallium (Ga), Tin (Sn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service