Skip to Content
Merck
All Photos(2)

Key Documents

34182

Supelco

Leucomalachite Green-d5

analytical standard

Synonym(s):

Bis-(4-dimethylaminophenyl)phenyl-d5-methane, LMG-D5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CH[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
335.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3/i5D,6D,7D,8D,9D

InChI key

WZKXBGJNNCGHIC-CFEWMVNUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Leucomalachite Green-d5 may be used as an internal standard for the determination of the triphenylmethane dye residues in rainbow trouts[1] by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muammer Kaplan et al.
Journal of chromatography. A, 1349, 37-43 (2014-05-29)
A rapid and simple LC-MS/MS method was developed and optimized for screening and confirmation of triphenylmethane dyes including malachite green (MG), leucomalachite green (LMG), crystal violet (CV), leucocrystal violet (LCV) and brilliant green (BG) in fish muscle with skin. Leucocrystal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service