IQ700XPKT1
Milli-Q® IQ Purification Kit
This product is replaced with IQ700XPKIT. Search to Explore!
Synonym(s):
Milli-Q® IQ 7003-5 Purification kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NB.85
Recommended Products
Quality Level
feature
Kit contains IPAK Gard® 3-5 Pretreatment Pack, IPAK Quanta® Polishing Cartridge and a Tank Vent Filter
packaging
pkg of 1 kit
compatibility
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
Looking for similar products? Visit Product Comparison Guide
General description
This water purification kit includes the following:
- IPAK Gard® 3-5 Pretreatment Pack efficiently removes colloids, particles and free chlorine from tap water to protect the reverse osmosis (RO) membrane and improve system performance.
- IPAK Quanta® polishing cartridge contains innovative IQnano™ ion-exchange media which achieves ion removal down to trace levels. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
- The Tank Vent Filter, redesigned for seamless integration, provides improved protection against airborne contaminants.
- This kit is designed for use with Milli-Q® IQ 7003-5 systems for the production of pure (Type 2) and ultrapure (Type 1) water.
Application
For optimal protection of the RO membrane, trace ion and organics removal, and advanced protection against airborne contaminants.
Features and Benefits
IPAK Gard® 3-5 Pretreatment Pack contains:
Tank Vent Filter contains:
IPAK Quanta® cartridge contains:
In addition, 63% of the parts suppliers for the IPAK Gard® cartridge and 78% of the parts suppliers for IPAK Quanta® cartridge included in this kit are ISO 14001 environmental management system-certified.
- Activated carbon to protect against free chlorine.
- Depth filter to protect against particulates and colloids.
Tank Vent Filter contains:
- Activated carbon to remove volatile organics.
- Soda lime to remove CO2.
- Membrane filter to prevent particulates and bacteria from entering the tank.
IPAK Quanta® cartridge contains:
- Innovative IQnano™ ion-exchange media to remove ions down to trace levels.
- High-grade synthetic activated carbon targets trace organic contaminants.
In addition, 63% of the parts suppliers for the IPAK Gard® cartridge and 78% of the parts suppliers for IPAK Quanta® cartridge included in this kit are ISO 14001 environmental management system-certified.
Legal Information
IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service