8.14160
2,5-Piperazinedione
for synthesis
Synonym(s):
2,5-Piperazinedione, Glycinanhydride, 2,5-Diketopiperazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
MDL number:
UNSPSC Code:
12352115
EC Index Number:
203-411-5
NACRES:
NA.22
Quality Level
form
powder
solubility
142 g/L
storage temp.
2-30°C
SMILES string
N1CC(=O)NCC1=O
InChI
1S/C4H6N2O2/c7-3-1-5-4(8)2-6-3/h1-2H2,(H,5,8)(H,6,7)
InChI key
BXRNXXXXHLBUKK-UHFFFAOYSA-N
Analysis Note
Assay (ex N): ≥ 98.0 %
Identity (IR): passes test
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service