Skip to Content
Merck
All Photos(1)

Key Documents

344101

Sigma-Aldrich

FSB

A fluorine analog of the amyloidophilic fluorescent probe BSB that crosses the blood-brain barrier and displays low toxicity.

Synonym(s):

FSB, (E,E)-1-Fluoro-2,5- bis(3-hydroxycarbonyl-4-hydroxy)styrylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17FO6
CAS Number:
Molecular Weight:
420.39
MDL number:
UNSPSC Code:
41105331
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1c(ccc(c1)\C=C\c3cc(c(cc3)O)C(=O)O)\C=C\c2cc(c(cc2)O)C(=O)O

InChI key

DZXZHGCGNRRJEG-WMWQKROPSA-N

General description

A fluorine analog of the amyloidophilic fluorescent probe BSB (Cat. No. 286895) that crosses the blood-brain barrier and displays low toxicity (LD50 >80 mg/kg). Suitable for non-invasive amyloid visualization in living transgenic Tg2576 mice by using 19F and 1H-MRI (Magnetic Resonance Imaging). Also useful for staining plaques and neurofibrillary tangles in brain tissue sections of Alzheimer′s diseased patients with improved sensitivity, and reduced background.
This fluorine analog of the amyloidophilic fluorescent probe BSB (Cat. No. 286895) crosses the blood-brain barrier and displays low toxicity (LD50 >80 mg/kg). Suitable for non-invasive amyloid visualization in living transgenic Tg2576 mice by using 19F and 1H-MRI. Also useful for staining plaques and neurofibrillary tangles in brain tissue sections of Alzheimer′s diseased patients, with improved sensitivity and reduced background.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Amyloidophilic fluorescent probe
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Higuchi, M., et al. 2005. Nat. Neurosci.8, 527.
Sato, K., et al. 2004. Eur. J. Med. Chem.39, 573.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service