Skip to Content
Merck
All Photos(1)

Key Documents

324473

Sigma-Aldrich

PARP Inhibitor IX, EB-47

The PARP Inhibitor IX, EB-47 controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor IX, EB-47

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27N9O6 · 2HCl · 2H2O
Molecular Weight:
646.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

potency

45 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, adenosine-substituted, isoindolinone compound that acts as a potent inhibitor of poly (ADP-ribose) polymerase-1 (PARP-1) (IC50 = 45 nM). Reported to offer cytoprotective effects against oxidative damage in cells and in in vivo models of reperfusion injury and inflammation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PARP-1
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jagtap, P.G., et al. 2004. Bioorg. Med. Chem. Lett.14, 81.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service