1.44198
Air Outlet Ring
for use with RCS® High Flow Touch
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
APEC® polycarbonate
Agency
non-compendial
feature
autoclavable
packaging
pkg of 1 kit
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with RCS High Flow Touch
storage temp.
room temp
General description
Air Outlet Ring is an additional accessory for use with RCS® microbial air sampler. The RCS® portable air sampler is used for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The Air Outlet Ring is made of autoclavable polycarbonate and is used to accelerate the outlet air.
Application
Air Outlet Ring is an additional accessory with the RCS® microbial air sampler used to accelerate the outlet air. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.
Compatibility
The Air Outlet Ring is compatible for use with RCS® Air Samplers of the High Flow series (RCS® High Flow Touch, RCS® High Flow) and RCS® Isolator.
Legal Information
APEC is a registered trademark of COVESTRO AG, Leverkusen, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service