Skip to Content
Merck
All Photos(1)

Documents

V-027

Supelco

(±)-O-Desmethylvenlafaxine-D6 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)−O-Desmethylvenlafaxine-D6 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19D6NO2
CAS Number:
Molecular Weight:
269.41
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC=C(C(CN(C([2H])([2H])[2H])C([2H])([2H])[2H])C2(O)CCCCC2)C=C1

InChI

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/i1D3,2D3

InChI key

KYYIDSXMWOZKMP-WFGJKAKNSA-N

General description

A certified reference solution suitable for use as an internal standard for quantitation of O-desmethylvenlafaxine levels in urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis. Also known as desvenlafaxine, O-desmethylvenlafaxine is an SNRI antidepressant marketed under the trade name Pristiq® for treatment of depression. O-desmethylvenlafaxine is also a major urinary metabolite of venlafaxine, an SNRI antidepressant sold as Effexor® or Efexor and used to treat major depressive disorder, generalized anxiety disorder, and other anxiety disorders associated with depression.

Application

  • Hydrodesulfurization research using pharmacosiderite Mo(4)P(3)O(16): This study utilized the microporous catalytic activity of synthesized Mo(4)P(3)O(16) under moderate temperatures for hydrodesulfurization, showing its potential in refining processes relevant to forensic toxicology for substance identification, including antidepressant drug metabolites (Ishikawa et al., 2024).
  • Synthesis of acyl cyclopentaquinolinones: Research demonstrated the construction of acyl cyclopentaquinolinones with potential implications for the synthesis and analysis of stable isotope-labeled compounds, helping to understand the biochemistry and metabolism of antidepressants (Xue et al., 2024).
  • Nutraceutical applications from biomass: The production and characterization of xylooligosaccharides from lignocellulosic biomass were reviewed, relevant for pharmacokinetic research by providing insights into plant-derived compound processing and applications, influencing stable isotope-labeled compound studies (Kumar et al., 2024).
  • Adverse event detection in pharmaceutical care: This study evaluated deep learning models for detecting adverse events from pharmaceutical care records, offering a methodological approach that could be adapted for monitoring effects related to antidepressant drugs in clinical settings (Nishioka et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Effexor is a registered trademark of American Home Product Corp.
Pristiq is a registered trademark of Wyeth LLC
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service