700324P
Avanti
Cholic acid 7-sulfate disodium salt
Avanti Research™ - A Croda Brand
Synonym(s):
Cholic acid 7-sulfate disodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H38Na2O8S
Molecular Weight:
532.60
UNSPSC Code:
12161700
NACRES:
NA.25
Assay
>99% (TLC)
Quality Level
form
powder
packaging
package of 1 × 1 mg
package of 1 × 5 mg
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
bile acids
shipped in
dry ice
storage temp.
−20°C
Packaging
5 mL Amber Glass Screw Cap Vial (700324-1MG)
5 mL Amber Glass Screw Cap Vial (700324-5MG)
5 mL Amber Glass Screw Cap Vial (700324-5MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service