Skip to Content
Merck
All Photos(1)

Key Documents

330729P

Avanti

C15 Glucosyl(β) Ceramide-d7 (d18:1-D7/15:0)

Avanti Research - A Croda Brand

Synonym(s):

D-glucosyl-β-1,1′-N-pentadecanoyl-D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68D7NO8
CAS Number:
Molecular Weight:
693.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (330729P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@@]([H])(NC(CCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

General description

C15 Glucosyl(β) Ceramide-d7 (d18:1-D7/15:0) is a deuterated derivative of C15 Glucosyl(β) Ceramide. β-Glucosyl ceramide (β-GlcCer) is an intracellular metabolite. It is the first intermediate in the synthesis of glycosphigolipids. β-GlcCer is an endogenous ligand of Mincle.

Biochem/physiol Actions

β-Glucosyl ceramide (β-GlcCer) exhibits immunostimulatory activity. Accumulation of β-GlcCer is associated with Gaucher disease.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular metabolite beta-glucosylceramide is an endogenous Mincle ligand possessing immunostimulatory activity
Nagata M, et al.
Proceedings of the National Academy of Sciences of the USA, 114(16), E3285-E3294 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service