Key Documents
Z689947
Sigma-Aldrich
Aldrich® column clamp
size S
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
Pricing and availability is not currently available.
size
S
L × diam.
211 mm × 11 mm , arm
jaw capacity
65-89 mm
Related Categories
General description
Corrosion resistant stainless steel. Ideal for holding chromatography columns and large cylindrical glassware. A large, flat thumbscrew/worm drive permits maximum tightening or removal in minimum time.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)