Key Documents
Z158399
Ace jacketed short-path distillation apparatus
Still body, with Vigreux indents
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Ace Glass 931760
Related Categories
General description
For small quantities. Jacketed still body, with 10/18 thermometer joint, one-piece 14/20 cow receiver, and 10 mL distillation flask with 14/20 top opening. Order thermometer adapter Z125350 and thermometer separately.
Replacement parts:
Z158364, Jacketed still body, plain
Z158372, Jacketed still body, with Vigreux indents
Choose from one of the most recent versions:
Certificates of Analysis (COA)