Key Documents
Z106429
Supelco
Black plastic caps
Solid top, PTFE-faced rubber liner, thread for 13-425, pkg of 200 ea
Synonym(s):
Protective Plastic Caps
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE-faced rubber liner
black phenolic cap
feature
closure type black phenolic solid top cap
packaging
pkg of 200 ea
fitting
thread for 13-425
General description
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
These autoclavable caps are designed to ensure the ultimate in product safety.
Choose from one of the most recent versions:
Certificates of Analysis (COA)